Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTAGTAAGCTCTTGCTTCATGTC[A/G]TTCAAGGGTCTTCCTTGTTTGCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615223 MIM: 616356 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFNE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFNE - interferon epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176891.4 | 1201 | Silent Mutation | AAC,AAT | N,N 194 | NP_795372.1 |
MIR31HG - MIR31 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |