Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGATGTAAAGGAAAGAATGTTAGA[C/G]TTCAAGCACAAGAAAATGCCCATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZCCHC7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZCCHC7 - zinc finger CCHC-type containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289119.1 | 501 | Missense Mutation | CTT,GTT | L,V 110 | NP_001276048.1 | |
NM_001289120.1 | 501 | Missense Mutation | CTT,GTT | L,V 110 | NP_001276049.1 | |
NM_001289121.1 | 501 | Missense Mutation | CTT,GTT | L,V 110 | NP_001276050.1 | |
NM_032226.2 | 501 | Missense Mutation | CTT,GTT | L,V 110 | NP_115602.2 | |
XM_005251608.4 | 501 | Missense Mutation | CTT,GTT | L,V 110 | XP_005251665.1 | |
XM_005251612.2 | 501 | Missense Mutation | CTT,GTT | L,V 110 | XP_005251669.1 | |
XM_011518050.1 | 501 | Missense Mutation | CTT,GTT | L,V 110 | XP_011516352.1 | |
XM_011518051.2 | 501 | Intron | XP_011516353.1 | |||
XM_017015197.1 | 501 | Missense Mutation | CTT,GTT | L,V 110 | XP_016870686.1 | |
XM_017015198.1 | 501 | Missense Mutation | CTT,GTT | L,V 110 | XP_016870687.1 | |
XM_017015199.1 | 501 | UTR 5 | XP_016870688.1 | |||
XM_017015200.1 | 501 | UTR 5 | XP_016870689.1 | |||
XM_017015201.1 | 501 | Intron | XP_016870690.1 |