Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTGTTGGTAGGCCAGAAGCGGA[C/T]TCCTAGAAGAGGGGAGCAGCAAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HABP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HABP4 - hyaluronan binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014282.3 | 318 | Missense Mutation | ACT,ATT | T,I 121 | NP_055097.2 | |
XM_005251812.3 | 318 | Missense Mutation | ACT,ATT | T,I 11 | XP_005251869.1 | |
XM_017014484.1 | 318 | UTR 5 | XP_016869973.1 |