Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAATCTACTTTGCCTTTGGCTAT[A/G]TTAATATTAGACAAACATTTCTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609198 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTSL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTSL1 - ADAMTS like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040272.5 | Intron | NP_001035362.3 | ||||
NM_052866.4 | Intron | NP_443098.3 | ||||
XM_011518063.2 | Intron | XP_011516365.1 | ||||
XM_011518064.2 | Intron | XP_011516366.1 | ||||
XM_011518067.1 | Intron | XP_011516369.1 | ||||
XM_011518068.2 | Intron | XP_011516370.1 | ||||
XM_011518070.2 | Intron | XP_011516372.1 | ||||
XM_017015310.1 | Intron | XP_016870799.1 | ||||
XM_017015311.1 | Intron | XP_016870800.1 | ||||
XM_017015312.1 | Intron | XP_016870801.1 | ||||
XM_017015313.1 | Intron | XP_016870802.1 | ||||
XM_017015314.1 | Intron | XP_016870803.1 |