Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGAAATACTTGTGGATGACATG[C/T]TGACTGTCGATGAATGGCTGGCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603564 MIM: 610891 MIM: 612865 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPM2 - dolichyl-phosphate mannosyltransferase polypeptide 2, regulatory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003863.3 | 775 | Silent Mutation | CAA,CAG | Q,Q 37 | NP_003854.1 |
FAM102A - family with sequence similarity 102 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIP5KL1 - phosphatidylinositol-4-phosphate 5-kinase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |