Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAATGTACAGATGCCCTCTACCA[G/A]AAATTAGACTTTACAGCACATCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616290 | ||||||||||||||||||||
Literature Links: |
ZNF658 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF658 - zinc finger protein 658 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317916.1 | Intron | NP_001304845.1 | ||||
NM_033160.6 | Intron | NP_149350.3 | ||||
XM_005272515.4 | Intron | XP_005272572.1 | ||||
XM_011545679.2 | Intron | XP_011543981.1 | ||||
XM_017014614.1 | Intron | XP_016870103.1 |