Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATCAGGTATGTGATTTGTGAGA[A/G]TGAGTCCTGGCCAGTTGTGAGAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607009 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPM6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRPM6 - transient receptor potential cation channel subfamily M member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177310.1 | Intron | NP_001170781.1 | ||||
NM_001177311.1 | Intron | NP_001170782.1 | ||||
NM_017662.4 | Intron | NP_060132.3 | ||||
XM_011518244.1 | Intron | XP_011516546.1 | ||||
XM_011518245.1 | Intron | XP_011516547.1 | ||||
XM_011518246.1 | Intron | XP_011516548.1 | ||||
XM_011518247.1 | Intron | XP_011516549.1 | ||||
XM_011518248.1 | Intron | XP_011516550.1 | ||||
XM_011518249.1 | Intron | XP_011516551.1 | ||||
XM_011518250.1 | Intron | XP_011516552.1 | ||||
XM_011518251.2 | Intron | XP_011516553.1 | ||||
XM_011518252.2 | Intron | XP_011516554.1 | ||||
XM_011518255.2 | Intron | XP_011516557.1 | ||||
XM_017014287.1 | Intron | XP_016869776.1 | ||||
XM_017014288.1 | Intron | XP_016869777.1 | ||||
XM_017014289.1 | Intron | XP_016869778.1 |