Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATAATCTAAAAATACAAACTTTT[A/C]ACTATTTTAAGACTAACAGATTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137350 MIM: 133090 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSN - gelsolin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STOM - stomatin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270526.1 | 1107 | UTR 3 | NP_001257455.1 | |||
NM_001270527.1 | 1107 | Intron | NP_001257456.1 | |||
NM_004099.5 | 1107 | UTR 3 | NP_004090.4 | |||
NM_198194.2 | 1107 | UTR 3 | NP_937837.1 |