Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCTTGTTGAGCTCCTCTAGGTC[A/G]CCCAGGTCGAGCTGGATGGAGTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605729 MIM: 138610 | ||||||||||||||||||||
Literature Links: |
AKNA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKNA - AT-hook transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317950.1 | 3984 | Silent Mutation | GGC,GGT | G,G 1393 | NP_001304879.1 | |
NM_001317952.1 | 3984 | Silent Mutation | GGC,GGT | G,G 1274 | NP_001304881.1 | |
NM_030767.5 | 3984 | Silent Mutation | GGC,GGT | G,G 1393 | NP_110394.3 | |
XM_005252244.2 | 3984 | Silent Mutation | GGC,GGT | G,G 1393 | XP_005252301.1 | |
XM_005252245.1 | 3984 | Silent Mutation | GGC,GGT | G,G 1393 | XP_005252302.1 | |
XM_005252247.4 | 3984 | Silent Mutation | GGC,GGT | G,G 1393 | XP_005252304.1 | |
XM_005252248.2 | 3984 | Silent Mutation | GGC,GGT | G,G 1361 | XP_005252305.1 | |
XM_006717294.1 | 3984 | Silent Mutation | GGC,GGT | G,G 1393 | XP_006717357.1 | |
XM_006717295.2 | 3984 | Intron | XP_006717358.1 | |||
XM_011519063.2 | 3984 | Silent Mutation | GGC,GGT | G,G 1274 | XP_011517365.1 | |
XM_011519064.2 | 3984 | Intron | XP_011517366.1 | |||
XM_011519065.2 | 3984 | Silent Mutation | GGC,GGT | G,G 1274 | XP_011517367.2 | |
XM_011519066.2 | 3984 | Intron | XP_011517368.1 | |||
XM_017015172.1 | 3984 | Silent Mutation | GGC,GGT | G,G 1274 | XP_016870661.1 |
ORM2 - orosomucoid 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |