Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCAAGAGCCGCTGGGTCATGGT[C/G]TCAGGGCCTGCGTCCACATACAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 103000 | ||||||||||||||||||||
Literature Links: |
AK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AK1 - adenylate kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000476.2 | 869 | Missense Mutation | GAC,GAG | D,E 123 | NP_000467.1 | |
NM_001318121.1 | 869 | Missense Mutation | GAC,GAG | D,E 123 | NP_001305050.1 | |
NM_001318122.1 | 869 | Missense Mutation | GAC,GAG | D,E 139 | NP_001305051.1 | |
XM_017014427.1 | 869 | Missense Mutation | GAC,GAG | D,E 123 | XP_016869916.1 | |
XM_017014428.1 | 869 | Missense Mutation | GAC,GAG | D,E 123 | XP_016869917.1 |
MIR4672 - microRNA 4672 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |