Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185640 MIM: 185620 MIM: 185630 MIM: 185660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPL7A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPL7A - ribosomal protein L7a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD36A - small nucleolar RNA, C/D box 36A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD36C - small nucleolar RNA, C/D box 36C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SURF1 - surfeit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SURF2 - surfeit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278928.1 | 638 | Missense Mutation | AAA,AGA | K,R 197 | NP_001265857.1 | |
NM_017503.4 | 638 | Missense Mutation | AAA,AGA | K,R 197 | NP_059973.4 |
SURF4 - surfeit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |