Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAGCAGCCCGGGAAGCCTTCCG[C/T]CTGGGGTCCCCATGGCGCCGGATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 100670 | ||||||||||||||||||||
Literature Links: |
ALDH1B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH1B1 - aldehyde dehydrogenase 1 family member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000692.4 | 417 | Silent Mutation | CGC,CGT | R,R 88 | NP_000683.3 | |
XM_011517802.2 | 417 | Silent Mutation | CGC,CGT | R,R 88 | XP_011516104.1 |