Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAATGGCATTGTCAATGAAAAT[A/G]TTGTTCCAGAAGGTTATGATTTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605978 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS13A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VPS13A - vacuolar protein sorting 13 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018037.1 | 990 | Missense Mutation | ATT,GTT | I,V 244 | NP_001018047.1 | |
NM_001018038.2 | 990 | Missense Mutation | ATT,GTT | I,V 244 | NP_001018048.1 | |
NM_015186.3 | 990 | Missense Mutation | ATT,GTT | I,V 244 | NP_056001.1 | |
NM_033305.2 | 990 | Missense Mutation | ATT,GTT | I,V 244 | NP_150648.2 |