Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGAAGATGAAGAGGCACTGCAT[C/T]ATTTGACCAGAGTTGAAGTGACAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610714 MIM: 600960 | ||||||||||||||||||||
Literature Links: |
PKN3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PKN3 - protein kinase N3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SET - SET nuclear proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122821.1 | 812 | Missense Mutation | CAT,TAT | H,Y 119 | NP_001116293.1 | |
NM_001248000.1 | 812 | Intron | NP_001234929.1 | |||
NM_001248001.1 | 812 | Intron | NP_001234930.1 | |||
NM_003011.3 | 812 | Missense Mutation | CAT,TAT | H,Y 106 | NP_003002.2 | |
XM_017015013.1 | 812 | Missense Mutation | CAT,TAT | H,Y 119 | XP_016870502.1 | |
XM_017015014.1 | 812 | Intron | XP_016870503.1 | |||
XM_017015015.1 | 812 | Intron | XP_016870504.1 | |||
XM_017015016.1 | 812 | Missense Mutation | CAT,TAT | H,Y 106 | XP_016870505.1 |