Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGATAAGTGGGTGATGGATGGTG[A/G]AAAGTCCTGAGAATGGGGAAAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605001 MIM: 614444 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANGPTL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANGPTL2 - angiopoietin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012098.2 | 2705 | UTR 3 | NP_036230.1 | |||
XM_006717030.3 | 2705 | Intron | XP_006717093.1 |