Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTGTGCCTTCAGTGGAAATCAA[C/T]AGTCATTGATTTTCAACAAATGTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRMO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRMO - tRNA methyltransferase O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016481.4 | Intron | NP_057565.3 | ||||
XM_005252047.3 | Intron | XP_005252104.1 | ||||
XM_005252048.4 | Intron | XP_005252105.1 | ||||
XM_005252049.4 | Intron | XP_005252106.1 | ||||
XM_005252050.4 | Intron | XP_005252107.1 | ||||
XM_011518776.2 | Intron | XP_011517078.1 | ||||
XM_011518777.1 | Intron | XP_011517079.1 |