Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGATGCAGCTGCTCCCAACTGAA[A/G]GGCTCCTGAAAACACTCGTGGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603505 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC14B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC14B - cell division cycle 14B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077181.1 | Intron | NP_001070649.1 | ||||
NM_003671.3 | Intron | NP_003662.1 | ||||
NM_033331.2 | Intron | NP_201588.1 | ||||
XM_005252288.3 | Intron | XP_005252345.1 | ||||
XM_011519147.2 | Intron | XP_011517449.1 | ||||
XM_011519148.2 | Intron | XP_011517450.1 | ||||
XM_011519149.2 | Intron | XP_011517451.1 | ||||
XM_011519150.2 | Intron | XP_011517452.1 | ||||
XM_011519151.2 | Intron | XP_011517453.1 | ||||
XM_011519152.2 | Intron | XP_011517454.1 | ||||
XM_011519153.2 | Intron | XP_011517455.1 | ||||
XM_011519156.2 | Intron | XP_011517458.2 | ||||
XM_011519158.1 | Intron | XP_011517460.1 | ||||
XM_011519159.2 | Intron | XP_011517461.2 | ||||
XM_017015240.1 | Intron | XP_016870729.1 | ||||
XM_017015241.1 | Intron | XP_016870730.1 | ||||
XM_017015242.1 | Intron | XP_016870731.1 | ||||
XM_017015243.1 | Intron | XP_016870732.1 | ||||
XM_017015244.1 | Intron | XP_016870733.1 | ||||
XM_017015245.1 | Intron | XP_016870734.1 | ||||
XM_017015246.1 | Intron | XP_016870735.1 | ||||
XM_017015247.1 | Intron | XP_016870736.1 | ||||
XM_017015248.1 | Intron | XP_016870737.1 | ||||
XM_017015249.1 | Intron | XP_016870738.1 |