Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCGCGGCGGCCCGCGCCCTTCG[A/C]AGTGGTCTTCGTGTCAGCCGACGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615299 | ||||||||||||||||||||
Literature Links: |
NXNL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NXNL2 - nucleoredoxin-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161625.1 | 531 | Missense Mutation | GAA,GCA | E,A 66 | NP_001155097.1 | |
NM_145283.2 | 531 | Missense Mutation | GAA,GCA | E,A 66 | NP_660326.2 | |
XM_005251727.3 | 531 | Missense Mutation | GAA,GCA | E,A 66 | XP_005251784.1 | |
XM_011518275.2 | 531 | Missense Mutation | GAA,GCA | E,A 66 | XP_011516577.1 | |
XM_011518276.2 | 531 | Missense Mutation | GAA,GCA | E,A 66 | XP_011516578.1 |