Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTCAAACAGCACTCACATAACA[C/T]GGAATTTATGCTGAAAATCCCATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604785 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTNNAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTNNAL1 - catenin alpha like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM245 - transmembrane protein 245 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032012.3 | 5542 | UTR 3 | NP_114401.2 | |||
XM_011518446.2 | 5542 | UTR 3 | XP_011516748.1 | |||
XM_011518447.1 | 5542 | UTR 3 | XP_011516749.1 | |||
XM_011518449.1 | 5542 | UTR 3 | XP_011516751.1 | |||
XM_011518451.1 | 5542 | UTR 3 | XP_011516753.1 | |||
XM_011518452.1 | 5542 | UTR 3 | XP_011516754.1 | |||
XM_017014571.1 | 5542 | UTR 3 | XP_016870060.1 | |||
XM_017014572.1 | 5542 | UTR 3 | XP_016870061.1 |