Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATACCGAATACAGTTTCTCACCA[C/T]AATGTGATCACTAGAAGTAAATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCAF10 - DDB1 and CUL4 associated factor 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286810.1 | Intron | NP_001273739.1 | ||||
NM_024345.4 | Intron | NP_077321.3 | ||||
XM_005251577.3 | Intron | XP_005251634.1 | ||||
XM_005251578.3 | Intron | XP_005251635.1 | ||||
XM_017015128.1 | Intron | XP_016870617.1 | ||||
XM_017015129.1 | Intron | XP_016870618.1 |
LOC105376037 - uncharacterized LOC105376037 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |