Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCTTTAAAAGCCTCTCTGACTC[C/T]TTATCCCAAATACCACATTACCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607141 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLIPR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLIPR2 - GLI pathogenesis related 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287010.1 | Intron | NP_001273939.1 | ||||
NM_001287011.1 | Intron | NP_001273940.1 | ||||
NM_001287012.1 | Intron | NP_001273941.1 | ||||
NM_001287013.1 | Intron | NP_001273942.1 | ||||
NM_001287014.1 | Intron | NP_001273943.1 | ||||
NM_022343.3 | Intron | NP_071738.1 |