Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTTCTGTCGGGCACAGAATATC[G/T]TTGCTCTTTTTTCAAAACTTGAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 100880 MIM: 609631 | ||||||||||||||||||||
Literature Links: |
ACO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACO1 - aconitase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DDX58 - DEXD/H-box helicase 58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014314.3 | 2740 | Missense Mutation | AAG,ACG | K,T 861 | NP_055129.2 |
LOC101060445 - uncharacterized LOC101060445 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |