Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGAACGAATAAAGTGTTTCATA[G/T]TTGGAAGTATGGTAAAGAGTCATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608871 MIM: 608872 | ||||||||||||||||||||
Literature Links: |
NIPSNAP3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NIPSNAP3A - nipsnap homolog 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NIPSNAP3B - nipsnap homolog 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018376.3 | 760 | Missense Mutation | AGT,ATT | S,I 87 | NP_060846.2 | |
XM_011518839.1 | 760 | Missense Mutation | AGT,ATT | S,I 87 | XP_011517141.1 | |
XM_017014889.1 | 760 | Missense Mutation | AGT,ATT | S,I 87 | XP_016870378.1 |