Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGACCCCCGGATGGGGCCGTCAG[C/T]GCCTCAGGCGGCAGGCGGCTCAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606073 | ||||||||||||||||||||
Literature Links: |
CYSRT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYSRT1 - cysteine rich tail 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDOR1 - NADPH dependent diflavin oxidoreductase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF208 - ring finger protein 208 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031297.4 | 765 | Silent Mutation | GCA,GCG | A,A 218 | NP_112587.2 |
RNF224 - ring finger protein 224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |