Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTCCCCTGTGTGAGTTCTCTGA[G/T]GTACTCTGAGATTGGATTTCTGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF782 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF782 - zinc finger protein 782 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001662.1 | 2470 | Missense Mutation | CAT,CCT | H,P 664 | NP_001001662.1 | |
XM_005251742.4 | 2470 | Missense Mutation | CAT,CCT | H,P 664 | XP_005251799.1 | |
XM_005251744.3 | 2470 | Missense Mutation | CAT,CCT | H,P 640 | XP_005251801.1 | |
XM_011518315.2 | 2470 | Missense Mutation | CAT,CCT | H,P 664 | XP_011516617.1 | |
XM_011518318.2 | 2470 | Missense Mutation | CAT,CCT | H,P 664 | XP_011516620.1 | |
XM_011518320.2 | 2470 | Missense Mutation | CAT,CCT | H,P 658 | XP_011516622.1 | |
XM_017014343.1 | 2470 | Missense Mutation | CAT,CCT | H,P 658 | XP_016869832.1 | |
XM_017014344.1 | 2470 | Missense Mutation | CAT,CCT | H,P 532 | XP_016869833.1 |