Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGCCTGGCTTCCTCTGGGGTGA[C/T]TTGGAAAAAAAAAGGAATTTTATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPDC1 - protein tyrosine phosphatase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253829.1 | Intron | NP_001240758.1 | ||||
NM_001253830.1 | Intron | NP_001240759.1 | ||||
NM_152422.4 | Intron | NP_689635.3 | ||||
NM_177995.2 | Intron | NP_818931.1 | ||||
XM_005251711.4 | Intron | XP_005251768.1 | ||||
XM_011518240.2 | Intron | XP_011516542.1 | ||||
XM_011518241.2 | Intron | XP_011516543.1 | ||||
XM_017014280.1 | Intron | XP_016869769.1 | ||||
XM_017014281.1 | Intron | XP_016869770.1 | ||||
XM_017014282.1 | Intron | XP_016869771.1 |