Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTCTTTAACGTAAGATTCCAAA[C/T]GACTCTGGAGATCTTCAGGTGGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602938 MIM: 611854 MIM: 603132 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAAT - bile acid-CoA:amino acid N-acyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL50 - mitochondrial ribosomal protein L50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019051.2 | Intron | NP_061924.1 |
ZNF189 - zinc finger protein 189 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |