Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTCTTCAGCAGTGTGACGGTAG[C/G]TTTTGCACAGGAAAGGTGGAGGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194552 | ||||||||||||||||||||
Literature Links: |
ZNF79 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF79 - zinc finger protein 79 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286696.1 | 658 | Missense Mutation | GCT,GGT | A,G 22 | NP_001273625.1 | |
NM_001286697.1 | 658 | Missense Mutation | GCT,GGT | A,G 22 | NP_001273626.1 | |
NM_001286698.1 | 658 | Intron | NP_001273627.1 | |||
NM_001322260.1 | 658 | Missense Mutation | GCT,GGT | A,G 22 | NP_001309189.1 | |
NM_007135.2 | 658 | Missense Mutation | GCT,GGT | A,G 46 | NP_009066.2 | |
XM_006717279.3 | 658 | Missense Mutation | GCT,GGT | A,G 22 | XP_006717342.1 |