Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCAGGACTGGTAAATAGCTTA[C/G]TCCATGATGGTTTAAAGTTTCCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605835 | ||||||||||||||||||||
Literature Links: |
TMEM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM2 - transmembrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135820.1 | 4355 | Missense Mutation | ACT,AGT | T,S 1272 | NP_001129292.1 | |
NM_013390.2 | 4355 | Missense Mutation | ACT,AGT | T,S 1335 | NP_037522.1 | |
XM_005251869.4 | 4355 | Missense Mutation | ACT,AGT | T,S 1335 | XP_005251926.1 |