Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGGGAAAACTTGCTTGGGTGCT[C/G]CAGGAGTCTGGATTTTAGTGCCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600777 | ||||||||||||||||||||
Literature Links: |
TTF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTF1 - transcription termination factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205296.1 | 3137 | Missense Mutation | GCA,GGA | A,G 339 | NP_001192225.1 | |
NM_007344.3 | 3137 | Missense Mutation | GCA,GGA | A,G 854 | NP_031370.2 | |
XM_006717273.3 | 3137 | Missense Mutation | GCA,GGA | A,G 854 | XP_006717336.2 | |
XM_017015105.1 | 3137 | Missense Mutation | GCA,GGA | A,G 887 | XP_016870594.1 |