Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTAATTTTCCGAGTGAATAGGAA[C/T]GCCTGAGTGGCCATTTAGAGTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611006 MIM: 613467 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISCA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ISCA1 - iron-sulfur cluster assembly 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZCCHC6 - zinc finger CCHC-type containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185059.1 | 5343 | UTR 3 | NP_001171988.1 | |||
NM_001185074.1 | 5343 | UTR 3 | NP_001172003.1 | |||
NM_024617.3 | 5343 | UTR 3 | NP_078893.2 | |||
XM_005252207.3 | 5343 | UTR 3 | XP_005252264.1 | |||
XM_005252208.2 | 5343 | Intron | XP_005252265.1 | |||
XM_006717283.1 | 5343 | UTR 3 | XP_006717346.1 | |||
XM_011519011.2 | 5343 | Intron | XP_011517313.1 | |||
XM_011519012.2 | 5343 | Intron | XP_011517314.1 | |||
XM_011519013.2 | 5343 | Intron | XP_011517315.1 | |||
XM_011519014.2 | 5343 | Intron | XP_011517316.1 | |||
XM_011519015.1 | 5343 | Intron | XP_011517317.1 | |||
XM_011519016.1 | 5343 | Intron | XP_011517318.1 | |||
XM_017015130.1 | 5343 | UTR 3 | XP_016870619.1 | |||
XM_017015131.1 | 5343 | UTR 3 | XP_016870620.1 | |||
XM_017015132.1 | 5343 | UTR 3 | XP_016870621.1 |