Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATTGCCATGGTAGCGCCCCAGCC[A/G]CAGCTTATAATACTCGCTCTCAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605001 MIM: 614444 | ||||||||||||||||||||
Literature Links: |
ANGPTL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANGPTL2 - angiopoietin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012098.2 | 1686 | Missense Mutation | CGG,TGG | R,W 396 | NP_036230.1 | |
XM_006717030.3 | 1686 | Intron | XP_006717093.1 |