Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAGCTGCAATTTACTCTTTTCT[C/G]TTTCCAGTATGTCCTCAGCTTCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PLGRKT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLGRKT - plasminogen receptor with a C-terminal lysine | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018465.3 | 628 | Missense Mutation | ACA,AGA | T,R 116 | NP_060935.2 | |
XM_005251510.4 | 628 | Missense Mutation | ACA,AGA | T,R 116 | XP_005251567.1 | |
XM_005251512.4 | 628 | Missense Mutation | ACA,AGA | T,R 83 | XP_005251569.1 | |
XM_011517960.2 | 628 | Missense Mutation | ACA,AGA | T,R 116 | XP_011516262.1 |