Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAGTACCGGGCCACCTGGCAGCG[A/C]CCACAGATGTTGAACTCCCGGAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613210 MIM: 611424 | ||||||||||||||||||||
Literature Links: |
DPH7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPH7 - diphthamide biosynthesis 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZMYND19 - zinc finger MYND-type containing 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138462.2 | 786 | Silent Mutation | GGG,GGT | G,G 188 | NP_612471.1 | |
XM_005266052.4 | 786 | Silent Mutation | GGG,GGT | G,G 249 | XP_005266109.1 | |
XM_017014255.1 | 786 | Intron | XP_016869744.1 |