Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCGCTGGGTCAACAGCTCCCGGA[G/T]CCTTCGGATCTCCTCCTGTTGCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602159 MIM: 606556 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CORO2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CORO2A - coronin 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003389.3 | 1755 | Missense Mutation | ATC,CTC | I,L 497 | NP_003380.3 | |
NM_052820.3 | 1755 | Missense Mutation | ATC,CTC | I,L 497 | NP_438171.1 | |
XM_011518986.2 | 1755 | Missense Mutation | ATC,CTC | I,L 497 | XP_011517288.1 |
TRIM14 - tripartite motif containing 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |