Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAAGAGAAGATGCTCACTTCAT[C/G]TATGGTTACCCCAAGAAGGGGCACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605513 | ||||||||||||||||||||
Literature Links: |
KIAA2026 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA2026 - KIAA2026 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017969.2 | 6777 | Intron | NP_001017969.2 | |||
XM_005251385.2 | 6777 | Intron | XP_005251442.1 | |||
XM_006716730.3 | 6777 | Intron | XP_006716793.1 | |||
XM_011517759.2 | 6777 | Intron | XP_011516061.1 | |||
XM_011517760.2 | 6777 | UTR 3 | XP_011516062.1 | |||
XM_011517761.2 | 6777 | Intron | XP_011516063.1 | |||
XM_017014331.1 | 6777 | Intron | XP_016869820.1 | |||
XM_017014332.1 | 6777 | Intron | XP_016869821.1 | |||
XM_017014333.1 | 6777 | Intron | XP_016869822.1 | |||
XM_017014334.1 | 6777 | Intron | XP_016869823.1 | |||
XM_017014335.1 | 6777 | Intron | XP_016869824.1 | |||
XM_017014336.1 | 6777 | Intron | XP_016869825.1 |
MLANA - melan-A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005511.1 | 6777 | Missense Mutation | ATC,ATG | I,M 9 | NP_005502.1 |