Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTCTTCATATTCAAAGTCTTCA[C/T]GGGGATGTCTTCTGTAATTTCTAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600709 | ||||||||||||||||||||
Literature Links: |
IARS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IARS - isoleucyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002161.5 | 4050 | Missense Mutation | ATG,GTG | M,V 1243 | NP_002152.2 | |
NM_013417.3 | 4050 | Missense Mutation | ATG,GTG | M,V 1243 | NP_038203.2 |