Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCATCAGGGCCTTCTCCTTTCC[A/G]GGTGCGACCACTGTGAAGCCAGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603308 | ||||||||||||||||||||
Literature Links: |
CTSV PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTSV - cathepsin V | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201575.1 | 940 | Intron | NP_001188504.1 | |||
NM_001333.3 | 940 | Silent Mutation | CCC,CCT | P,P 230 | NP_001324.2 |