Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACTGTGCCATTGTCCCACTTGA[C/G]CTTTTGGCTAGACTGGGTCTTTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613747 MIM: 609787 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF24 - kinesin family member 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194313.2 | 4206 | UTR 3 | NP_919289.2 | |||
XM_005251451.2 | 4206 | UTR 3 | XP_005251508.1 | |||
XM_006716764.2 | 4206 | UTR 3 | XP_006716827.1 | |||
XM_011517861.1 | 4206 | UTR 3 | XP_011516163.1 | |||
XM_011517863.2 | 4206 | UTR 3 | XP_011516165.1 | |||
XM_017014697.1 | 4206 | UTR 3 | XP_016870186.1 | |||
XM_017014698.1 | 4206 | UTR 3 | XP_016870187.1 |
UBAP1 - ubiquitin associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171201.1 | 4206 | Intron | NP_001164672.1 | |||
NM_001171202.1 | 4206 | Intron | NP_001164673.1 | |||
NM_001171203.2 | 4206 | Intron | NP_001164674.1 | |||
NM_001171204.2 | 4206 | Intron | NP_001164675.1 | |||
NM_016525.4 | 4206 | Intron | NP_057609.2 | |||
XM_006716779.3 | 4206 | Intron | XP_006716842.1 | |||
XM_011517898.2 | 4206 | Intron | XP_011516200.1 | |||
XM_011517899.1 | 4206 | Intron | XP_011516201.1 | |||
XM_011517900.1 | 4206 | Intron | XP_011516202.1 | |||
XM_017014801.1 | 4206 | Intron | XP_016870290.1 | |||
XM_017014802.1 | 4206 | Intron | XP_016870291.1 |