Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACAACGAAATTATAAAGCCTGC[C/T]AAATACTTCTCAGAATTGGAAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MSANTD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MSANTD3 - Myb/SANT DNA binding domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198805.1 | 63 | Silent Mutation | GCC,GCT | A,A 10 | NP_001185734.1 | |
NM_001198806.1 | 63 | Silent Mutation | GCC,GCT | A,A 10 | NP_001185735.1 | |
NM_001198807.1 | 63 | Silent Mutation | GCC,GCT | A,A 10 | NP_001185736.1 | |
NM_080655.2 | 63 | Silent Mutation | GCC,GCT | A,A 10 | NP_542386.1 |
MSANTD3-TMEFF1 - MSANTD3-TMEFF1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198812.1 | 63 | Silent Mutation | GCC,GCT | A,A 10 | NP_001185741.1 |