Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTTCTGGAGGACTTCAACTTG[C/T]TCTGAAATCTTTGTTTTCTCCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611253 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF27 - kinesin family member 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271927.1 | 3236 | Silent Mutation | NP_001258856.1 | |||
NM_001271928.1 | 3236 | Silent Mutation | NP_001258857.1 | |||
NM_017576.2 | 3236 | Silent Mutation | NP_060046.1 |