Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTTGTAACTTATTTGCTATGAC[C/T]TTGGGCAAATTACGGTTAGTTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600062 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD23B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAD23B - RAD23 homolog B, nucleotide excision repair protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244713.1 | 68 | UTR 5 | NP_001231642.1 | |||
NM_001244724.1 | 68 | Intron | NP_001231653.1 | |||
NM_002874.4 | 68 | Intron | NP_002865.1 |