Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGCTAAGTGACCCCAAGAAACG[C/G]GGCCTGTATGACCAGTATGGGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611328 | ||||||||||||||||||||
Literature Links: |
DNAJB5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJB5 - DnaJ heat shock protein family (Hsp40) member B5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135004.2 | 327 | Silent Mutation | CGC,CGG | R,R 175 | NP_001128476.2 | |
NM_001135005.2 | 327 | Silent Mutation | CGC,CGG | R,R 133 | NP_001128477.1 | |
NM_012266.5 | 327 | Silent Mutation | CGC,CGG | R,R 61 | NP_036398.3 | |
XM_005251428.3 | 327 | Silent Mutation | CGC,CGG | R,R 61 | XP_005251485.1 | |
XM_006716751.2 | 327 | Silent Mutation | CGC,CGG | R,R 61 | XP_006716814.1 | |
XM_011517842.2 | 327 | Silent Mutation | CGC,CGG | R,R 133 | XP_011516144.1 |
DNAJB5-AS1 - DNAJB5 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |