Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGCTGCGGGTGAGGAGGACGA[A/G]GGCGACATCGCGCTGCAGATCCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604949 | ||||||||||||||||||||
Literature Links: |
GADD45G PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GADD45G - growth arrest and DNA damage inducible gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006705.3 | 317 | Silent Mutation | GAA,GAG | E,E 69 | NP_006696.1 | |
XM_011518163.2 | 317 | Silent Mutation | GAA,GAG | E,E 35 | XP_011516465.1 |