Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCCCAGCAATTTCTTCATAGCAT[A/C]TTTGACTTCCTTGTTTCTTAAACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR2K2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR2K2 - olfactory receptor family 2 subfamily K member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_205859.1 | 892 | Missense Mutation | GAT,TAT | D,Y 298 | NP_995581.1 | |
XM_011518520.2 | 892 | Missense Mutation | GAT,TAT | D,Y 298 | XP_011516822.1 |