Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTGATGCCCTCTGAGATTTGA[C/T]TTCTGCCTGAATGTTTTTCCACACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF782 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF782 - zinc finger protein 782 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001662.1 | 2285 | Silent Mutation | AAA,AAG | K,K 602 | NP_001001662.1 | |
XM_005251742.4 | 2285 | Silent Mutation | AAA,AAG | K,K 602 | XP_005251799.1 | |
XM_005251744.3 | 2285 | Silent Mutation | AAA,AAG | K,K 578 | XP_005251801.1 | |
XM_011518315.2 | 2285 | Silent Mutation | AAA,AAG | K,K 602 | XP_011516617.1 | |
XM_011518318.2 | 2285 | Silent Mutation | AAA,AAG | K,K 602 | XP_011516620.1 | |
XM_011518320.2 | 2285 | Silent Mutation | AAA,AAG | K,K 596 | XP_011516622.1 | |
XM_017014343.1 | 2285 | Silent Mutation | AAA,AAG | K,K 596 | XP_016869832.1 | |
XM_017014344.1 | 2285 | Silent Mutation | AAA,AAG | K,K 470 | XP_016869833.1 |