Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAGGACCAGGGCTCCGCCTCCCC[A/G]GTCAGTGTGAGCAGCGATGACTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614056 | ||||||||||||||||||||
Literature Links: |
PPP1R26 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1R26 - protein phosphatase 1 regulatory subunit 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014811.3 | 1470 | Intron | CCA,CCG | P,P 213 | NP_055626.3 | |
XM_005263411.2 | 1470 | Intron | XP_005263468.1 | |||
XM_017015355.1 | 1470 | Silent Mutation | CCA,CCG | P,P 213 | XP_016870844.1 | |
XM_017015356.1 | 1470 | Silent Mutation | CCA,CCG | P,P 213 | XP_016870845.1 | |
XM_017015357.1 | 1470 | Silent Mutation | CCA,CCG | P,P 213 | XP_016870846.1 | |
XM_017015358.1 | 1470 | Silent Mutation | CCA,CCG | P,P 213 | XP_016870847.1 | |
XM_017015359.1 | 1470 | Silent Mutation | CCA,CCG | P,P 213 | XP_016870848.1 | |
XM_017015360.1 | 1470 | Silent Mutation | CCA,CCG | P,P 213 | XP_016870849.1 | |
XM_017015361.1 | 1470 | Silent Mutation | CCA,CCG | P,P 213 | XP_016870850.1 | |
XM_017015362.1 | 1470 | Silent Mutation | CCA,CCG | P,P 213 | XP_016870851.1 |