Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCATCGAGGCATGGGGGTCCACA[A/C]TGCAATGTTTTTGTGGAACATGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 189980 MIM: 602238 | ||||||||||||||||||||
Literature Links: |
ABL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABL1 - ABL proto-oncogene 1, non-receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005157.5 | 553 | Intron | NP_005148.2 | |||
NM_007313.2 | 553 | Missense Mutation | CAA,CAC | Q,H 38 | NP_009297.2 |
EXOSC2 - exosome component 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |