Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTACCTCAGCACCCGCACCCACGG[C/T]GCCGTCTTCAACCTCGAGTACTCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DCAF10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCAF10 - DDB1 and CUL4 associated factor 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286810.1 | 609 | Intron | NP_001273739.1 | |||
NM_024345.4 | 609 | Silent Mutation | GGC,GGT | G,G 169 | NP_077321.3 | |
XM_005251577.3 | 609 | Silent Mutation | GGC,GGT | G,G 169 | XP_005251634.1 | |
XM_005251578.3 | 609 | Silent Mutation | GGC,GGT | G,G 169 | XP_005251635.1 | |
XM_017015128.1 | 609 | Silent Mutation | GGC,GGT | G,G 169 | XP_016870617.1 | |
XM_017015129.1 | 609 | Intron | XP_016870618.1 |
LOC105376037 - uncharacterized LOC105376037 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |