Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCAGAAGTCATTCCGGAGAATA[C/T]TGACCTGGTGACTTTGGGGTATGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602348 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNAPC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNAPC3 - small nuclear RNA activating complex polypeptide 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039697.1 | 558 | Missense Mutation | ACT,ATT | T,I 125 | NP_001034786.1 | |
XM_017015056.1 | 558 | Missense Mutation | ACT,ATT | T,I 125 | XP_016870545.1 | |
XM_017015057.1 | 558 | Missense Mutation | ACT,ATT | T,I 125 | XP_016870546.1 |